Ethylvanillin 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Ethoxy-4-hydroxybenzaldehyde, 3-(Ethoxy-d5)-4-hydroxybenzaldehyde

15603-0050 15603-1000 15603-5000
TS15603-0050EA 24 USD
TS15603-0050 TS15603-1000 TS15603-5000
Ethylvanillin 97%
Ethylvanillin
Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 285
Melting Pt: 76…79 °C
Flash Pt: 100
Storage Temperature: Ambient
MDL Number: MFCD00006944
CAS Number: 121-32-4
EINECS: 204-464-7
Merck Index: 13,03890

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 75 to 79 °C
GC ≥99.75 %
Color scale ≤10 APHA
Residue after evaporation ≤0.0005 %
UV transmittance ≥95 % (at 250 nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR