Tetraethylene glycol 99.5%

Supplier: Thermo Scientific Chemicals

Synonyms: 2,2'-((Oxybis(ethane-2,1-diyl))bis(oxy))diethanol, Tetraglycol

14959-0010 14959-0025 14959-0100
TS14959-0010EA 67 USD
TS14959-0010 TS14959-0025 TS14959-0100
Tetraethylene glycol 99.5%
Tetraethylene glycol
Formula: C₈H₁₈O₅
Boiling Pt: 324…330 °C (760 mmHg)
Melting Pt: –4.1 °C
Density: 1.12 g/cm³
Flash Pt: 176 °C
Storage Temperature: Ambient
MDL Number: MFCD00002879
CAS Number: 112-60-7
EINECS: 203-989-9

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4580 to 1.4600 (20 °C, 589 nm)
GC ≥99.4 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR