α,α,α-Trifluoroacetophenone 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,2,2-Trifluoroacetophenone, Phenacyl trifluoride, Trifluoroacetylbenzene, 1-Phenyl-2,2,2-trifluoro-1-ethanone

14835-0050 14835-0250 14835-1000
TS14835-0050EA 46 USD
TS14835-0050 TS14835-0250 TS14835-1000
α,α,α-Trifluoroacetophenone 99%
α,α,α-Trifluoroacetophenone
Formula: C₈H₅F₃O
MW: 174.12 g/mol
Boiling Pt: 165
Melting Pt: -40
Density: 1.24 g/cm³
Flash Pt: 41 °C
Storage Temperature: Ambient
MDL Number: MFCD00000420
CAS Number: 434-45-7
EINECS: 207-103-1
UN: 1993
ADR: 3 III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4580 to 1.4600 (20 °C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR