2,6,10,14-Tetramethylpentadecane 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Pristane

13846-0050 13846-0500 13846-2500
TS13846-0050EA 42 USD
TS13846-0050 TS13846-0500 TS13846-2500
2,6,10,14-Tetramethylpentadecane 95%
2,6,10,14-Tetramethylpentadecane
Formula: C₁₉H₄₀
MW: 268.53 g/mol
Boiling Pt: 296
Density: 0.78 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00008952
CAS Number: 1921-70-6
EINECS: 217-650-8
Merck Index: 12,07932

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR