3,3-Dimethylbutanone 96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1,1,1-Trimethylacetone, Methyl tert-butyl ketone, Pinacolone, tert-Butyl methyl ketone, 3,3-Dimethyl-2-butanone

13125-0025 13125-1000 13125-5000
TS13125-0025EA 845 USD
TS13125-0025 TS13125-1000 TS13125-5000
3,3-Dimethylbutanone 96%
3,3-Dimethylbutanone
Formula: C₆H₁₂O
MW: 100.16 g/mol
Boiling Pt: 106
Melting Pt: -53
Density: 0.8 g/cm³
Flash Pt: 5 °C
Storage Temperature: Ambient
MDL Number: MFCD00008846
CAS Number: 75-97-8
EINECS: 200-920-4
UN: 1224
ADR: 3 II
Merck Index: 12,07594

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3950 to 1.3980 (20 °C, 589 nm)
GC ≥95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR