3-Hydroxybenzaldehyde 98.5%

Supplier: Thermo Scientific Chemicals
Warning

12096-1000 12096-5000
TS12096-1000EA 68 USD
TS12096-1000 TS12096-5000
3-Hydroxybenzaldehyde 98.5%
3-Hydroxybenzaldehyde
Formula: C₇H₆O₂
MW: 122.12 g/mol
Boiling Pt: 241
Melting Pt: 101…105 °C
Storage Temperature: Ambient
MDL Number: MFCD00003368
CAS Number: 100-83-4
EINECS: 202-892-9

Order Now

Specification Test Results

Appearance Light yellow to brown Crystalline powder
Infrared spectrum Conforms
Melting point 101 to 105 °C
GC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR