4-tert-Butylbenzenesulphonyl chloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
4-tert-Butylbenzene-1-sulfonyl chloride, 4-tert-Butylbenzene-1-sulphonyl chloride
30347-0100
30347-0500
TS30347-0100EA
54
USD
TS30347-0100
TS30347-0500
4-tert-Butylbenzenesulphonyl chloride 98%
4-tert-Butylbenzenesulphonyl chloride
Formula:
C₁₀H₁₃ClO₂S MW: 232.73 Boiling Pt: 165 Melting Pt: 83 Storage Temperature: Ambient |
MDL Number:
MFCD00007449 CAS Number: 15084-51-2 UN: 3261 ADR: 8 II |
Specification Test Results
Appearance | White to off-white to light yellow Powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Melting point | 78 to 84 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...