(S)-(+)-2,3,7,7a-Tetrahydro-7a-methyl-1H-indene-1,5(6H)-dione 90%

Supplier: Thermo Scientific Chemicals
Warning

29647-0010 29647-0050
TS29647-0010EA 61 USD
TS29647-0010 TS29647-0050
(S)-(+)-2,3,7,7a-Tetrahydro-7a-methyl-1H-indene-1,5(6H)-dione 90%
(S)-(+)-2,3,7,7a-Tetrahydro-7a-methyl-1H-indene-1,5(6H)-dione
Formula: C₁₀H₁₂O₂
Melting Pt: 55…64 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012272
CAS Number: 17553-86-5

Order Now

Specification Test Results

Appearance White to yellow to beige Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 55 to 64 °C
GC ≥88 %
Specific optical rotation +330° to +360° (20 °C, 589 nm) (c=1, toluene)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR