1,4-Dioxaspiro[4.5]decan-8-one 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,4-Cyclohexanedione monoethylene acetal, 1,4-Cyclohexanedione monoethylene ketal

23564-0250 23564-1000
TS23564-0250EA 73 USD
TS23564-0250 TS23564-1000
1,4-Dioxaspiro[4.5]decan-8-one 98%
1,4-Dioxaspiro[4.5]decan-8-one
Formula: C₈H₁₂O₃
MW: 156.18 g/mol
Melting Pt: 74…76 °C
Storage Temperature: Ambient
MDL Number: MFCD00010214
CAS Number: 4746-97-8

Order Now

Specification Test Results

Appearance White to beige Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 70 to 74 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR