1,7-Octadiyne 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Octa-1,7-diyne

17134-0100 17134-0500
TS17134-0100EA 95 USD
TS17134-0100 TS17134-0500
1,7-Octadiyne 98%
1,7-Octadiyne
Formula: C₈H₁₀
Boiling Pt: 136 °C (1013 hPa)
Melting Pt: –32 °C
Density: 0.8 g/cm³
Flash Pt: 23 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008580
CAS Number: 871-84-1
EINECS: 212-815-0
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4455 to 1.4485 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR