Quinidine, anhydrous 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (+)-Quinidine

16368-0250 16368-1000
TS16368-0250EA 211 USD
TS16368-0250 TS16368-1000
Quinidine, anhydrous 98%
Quinidine
Formula: C20H24N2O2
MW: 324.42 g/mol
Melting Pt: 170…175 °C
Storage Temperature: Ambient
MDL Number: MFCD00135581
CAS Number: 56-54-2
UN: 1544
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 170 to 175 °C
Additional info Remainder dihydroquinidine
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR