2-Bromothiophene 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Thienyl bromide

15429-0010 15429-0500 15429-2500
TS15429-0010EA 478 USD
TS15429-0010 TS15429-0500 TS15429-2500
2-Bromothiophene 98%
2-Bromothiophene
Formula: C₄H₃BrS
Boiling Pt: 149…151 °C (760 mmHg)
Melting Pt: –10 °C
Density: 1.68 g/cm³
Flash Pt: 52 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005417
CAS Number: 1003-09-4
EINECS: 213-699-4
UN: 2929
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear colorless to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.5850 to 1.5870 (20°C, 589 nm)
GC ≥97.5 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR