Tropine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: endo-8-Methyl-8-azabicyclo[3.2.1]octan-3-ol

14056-0250 14056-1000
TS14056-0250EA 98 USD
TS14056-0250 TS14056-1000
Tropine 98%
Tropine
Formula: C₈H₁₅NO
MW: 141.21 g/mol
Boiling Pt: 233 °C (760 mmHg)
Melting Pt: 62…64 °C
Density: 1.04 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00180667
CAS Number: 120-29-6
EINECS: 204-384-2

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Water ≤2 % (K.F.)
Titration with HClO4 ≥97.5 % (On dried substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR