Tetraphenylcyclopentadienone 99%

Supplier: Thermo Scientific Chemicals

13854-0050 13854-0250 13854-1000
TS13854-0050EA 41 USD
TS13854-0050 TS13854-0250 TS13854-1000
Tetraphenylcyclopentadienone 99%
Tetraphenylcyclopentadienone
Formula: C₂₉H₂₀O
Melting Pt: 217…222 °C
Storage Temperature: Ambient
MDL Number: MFCD00001407
CAS Number: 479-33-4
EINECS: 207-530-3

Order Now

Specification Test Results

Appearance Black Crystalline powder
Infrared spectrum Conforms
Melting point 217 to 222 °C
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR