7-Hydroxycoumarin 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Umbelliferone, 7-Hydroxy-2H-chromen-2-one, 7-Hydroxy-2-benzopyrone

12111-0250 12111-1000
TS12111-0250EA 76 USD
TS12111-0250 TS12111-1000
7-Hydroxycoumarin 99%
7-Hydroxycoumarin
Formula: C₉H₆O₃
MW: 162.14 g/mol
Boiling Pt: 382
Melting Pt: 228…234 °C
Storage Temperature: Ambient
MDL Number: MFCD00006878
CAS Number: 93-35-6
EINECS: 202-240-3
Merck Index: 12,09981

Order Now

Specification Test Results

Appearance White to yellow to beige to light brown Powder or crystals and/or agglomerates
Infrared spectrum Conforms
Melting point 228 to 234 °C
HPLC ≥98.5 %
Titration with NaOH 98.5 to 101.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR