2,6-Dichloroaniline 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
11310-0250
11310-1000
11310-5000
TS11310-0250EA
36
USD
TS11310-0250
TS11310-1000
TS11310-5000
2,6-Dichloroaniline 98%
2,6-Dichloroaniline
Formula:
C₆H₅Cl₂N Boiling Pt: 228 °C (760 mmHg) Melting Pt: 35…39 °C Flash Pt: 118 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007675 CAS Number: 608-31-1 EINECS: 210-160-5 UN: 3442 ADR: 6.1,II |
Specification Test Results
Appearance | White to brown Crystalline solid (May darken on storage) |
Infrared spectrum | Conforms |
Melting point | 35 to 39 °C |
GC | ≥97.5 % |
BSC Certification | Passes |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...