Dibutyl disulphide 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Dibutyldisulphide, n-Butyl disulfide, n-Butyl disulphide, Dibutyldisulfide

10792-0250 10792-1000 10792-5000
TS10792-0250EA 47 USD
TS10792-0250 TS10792-1000 TS10792-5000
Dibutyl disulphide 97%
Dibutyl disulphide
Formula: C₈H₁₈S₂
MW: 178.36 g/mol
Boiling Pt: 229…233 °C (760 mmHg)
Density: 0.938 g/cm³
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00009467
CAS Number: 629-45-8
EINECS: 211-091-3

Order Now

Specification Test Results

Appearance Clear colorless to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.4915 to 1.4935 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR