2,2-Dimethylcyclopentanone 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,2-Dimethylcyclopentan-1-one

39940-0010 39940-0050
TS39940-0010EA 62 USD
TS39940-0010 TS39940-0050
2,2-Dimethylcyclopentanone 97%
2,2-Dimethylcyclopentanone
Formula: C₇H₁₂O
Boiling Pt: 143…145 °C (760 mmHg)
Density: 0.89 g/cm³
Flash Pt: 32 °C
Storage Temperature: Ambient
MDL Number: MFCD00049164
CAS Number: 4541-32-6
UN: 1224
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4320 to 1.4340 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR