2-(3-Trifluoromethylphenyl)ethylamine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-(Trifluoromethyl)phenethylamine

36552-0010 36552-0050
TS36552-0010EA 24 USD
TS36552-0010 TS36552-0050
2-(3-Trifluoromethylphenyl)ethylamine 98%
2-(3-Trifluoromethylphenyl)ethylamine
Formula: C₉H₁₀F₃N
MW: 189.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00040756
CAS Number: 52516-30-0
UN: 2735
ADR: 8 III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4635 to 1.4655 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR