Bis(2-oxo-3-oxazolidinyl)phosphinic chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: BOP-Cl

31006-0050 31006-0250
TS31006-0050EA 68 USD
TS31006-0050 TS31006-0250
Bis(2-oxo-3-oxazolidinyl)phosphinic chloride 97%
Bis(2-oxo-3-oxazolidinyl)phosphinic chloride
Formula: C₆H₈ClN₂O₅P
Melting Pt: 191 °C
Storage Temperature: Ambient
MDL Number: MFCD00010077
CAS Number: 68641-49-6
UN: 1759
ADR: 8,II

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR