Scopoletin (7-hydroxy-6-methoxycumarin) 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Scopoletin, 7-Hydroxy-6-methoxy-2H-chromen-2-one
30290-0010
30290-0500
30290-2500
TS30290-0010EA
762
USD
TS30290-0010
TS30290-0500
TS30290-2500
Scopoletin (7-hydroxy-6-methoxycumarin) 95%
Scopoletin (7-Hydroxy-6-methoxycumarin)
Formula:
C₁₀H₈O₄ MW: 192.1681 g/mol Melting Pt: 200…207 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006872 CAS Number: 92-61-5 EINECS: 202-171-9 |
Specification Test Results
Appearance | Light yellow to beige Powder |
Infrared spectrum | Conforms |
Melting point | 200 to 207 °C |
HPLC | ≥94.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...