Phloretin 98%
Supplier: Thermo Scientific Chemicals
30765-1000
30765-5000
TS30765-1000EA
98
USD
TS30765-1000
TS30765-5000
Phloretin 98%
Phloretin
Formula:
C₁₅H₁₄O₅ MW: 274.28 g/mol Melting Pt: 260…262 °C Storage Temperature: Freezer |
MDL Number:
MFCD00002288 CAS Number: 60-82-2 EINECS: 200-488-7 |
Specification Test Results
Appearance | White to beige Powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...