Diphenyliodonium hexafluorophosphate 97%

Supplier: Thermo Scientific Chemicals
Danger

45091-0050 45091-0250 45091-1000
TS45091-0050EA 64 USD
TS45091-0050 TS45091-0250 TS45091-1000
Diphenyliodonium hexafluorophosphate 97%
Diphenyliodonium hexafluorophosphate
Formula: C₁₂H₁₀F₆IP
Melting Pt: 140…144 °C
Storage Temperature: Ambient
MDL Number: MFCD00061398
CAS Number: 58109-40-3
UN: 1759
ADR: 8,II

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 138 to 143 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR