Sodium dodecylbenzenesulphonate
Supplier: Thermo Scientific Chemicals
|
Danger
|
32591-0010
32591-0050
32591-0250
TS32591-0010EA
49
USD
TS32591-0010
TS32591-0050
TS32591-0250
Sodium dodecylbenzenesulphonate
Sodium dodecylbenzenesulphonate
Formula:
C₁₈H₂₉NaO₃S Melting Pt: > 300 °C Density: 1 g/cm³ (20 °C) |
MDL Number:
MFCD00011508 CAS Number: 25155-30-0 EINECS: 246-680-4 Merck Index: 1308686 |
Specification Test Results
Appearance | White to yellow Crystalline powder |
Infrared spectrum | Conforms |
Assay | ≥83 % |
Water | ≤2 % |
pH | 6.5 to 9.5 (1% soln.) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...