(±)-3-Bromocyclohexene 95%

Supplier: Thermo Scientific Chemicals
Warning

33512-0100 33512-0500 33512-2500
TS33512-0100EA 81 USD
TS33512-0100 TS33512-0500 TS33512-2500
(±)-3-Bromocyclohexene 95%
(±)-3-Bromocyclohexene
Formula: C₆H₉Br
Boiling Pt: 64…65 °C (15 mmHg)
Density: 1.38 g/cm³
Flash Pt: 54 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00013775
CAS Number: 1521-51-3
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow to light brown Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5280 to 1.5320 (20°C, 589 nm)
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR