2,2,2-Tribromoethanol 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tribromoethyl alcohol

42143-0500 42143-0100 42143-2500
TS42143-0500EA 227 USD
TS42143-0500 TS42143-0100 TS42143-2500
2,2,2-Tribromoethanol 99%
2,2,2-Tribromoethanol
Formula: Br₃CCH₂OH
Melting Pt: 73…79 °C
Storage Temperature: Ambient
MDL Number: MFCD00004671
CAS Number: 75-80-9
EINECS: 200-903-1
Merck Index: 1209743

Order Now

Specification Test Results

Appearance White to yellow to light brown Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 73 to 82 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR