1,6-Heptadiyne 98%

Supplier: Thermo Scientific Chemicals
Danger

30942-0010 30942-0050
TS30942-0010EA 51 USD
TS30942-0010 TS30942-0050
1,6-Heptadiyne 98%
1,6-Heptadiyne
Formula: C₇H₈
MW: 92.14 g/mol
Boiling Pt: 108
Melting Pt: -85
Density: 0.805 g/cm³
Flash Pt: 9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00014925
CAS Number: 2396-63-6
EINECS: 219-253-5
UN: 1993
ADR: 3 II

Order Now

Specification Test Results

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4400 to 1.4440 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR