Bromoacetonitrile 97%

Supplier: Thermo Scientific Chemicals
Danger

22186-0050 22186-0250 22186-1000
TS22186-0050EA 40 USD
TS22186-0050 TS22186-0250 TS22186-1000
Bromoacetonitrile 97%
Bromoacetonitrile
Formula: BrCNCH₂
MW: 119.95 g/mol
Boiling Pt: 150
Melting Pt: -36
Density: 1.72 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00001884
CAS Number: 590-17-0
EINECS: 209-672-1
UN: 2922
ADR: 8 II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4780 to 1.4810 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR