Triformamide 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: trialkanoylamines, N,N-Diformylformamide, Formamide, N,N-diformyl-

38044-0050 380440010
TS38044-0050EA 104 USD
TS38044-0050 TS380440010
Triformamide 97%
Triformamide
Formula: C₃H₃NO₃
MW: 101.06 g/mol
Melting Pt: 100…101 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08059356
CAS Number: 25891-31-0

Order Now

Specification Test Results

Appearance White Crystals
Infrared spectrum Conforms
Melting point 95.0 to 100.0 °C
Elemental Analysis ≥96.0 % (ex C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR