2-Acetamido-6-hydroxypurine 99%

Supplier: Thermo Scientific Chemicals

25840-0010 25840-0050
TS25840-0010EA 131 USD
TS25840-0010 TS25840-0050
2-Acetamido-6-hydroxypurine 99%
2-Acetamido-6-hydroxypurine
Formula: C₇H₇N₅O₂
Melting Pt: >260 °C
Storage Temperature: Ambient
MDL Number: MFCD00078201
CAS Number: 19962-37-9

Order Now

Specification Test Results

Appearance White to beige Powder
Infrared spectrum Conforms
Titration with HClO4 ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR