Benzo[b]furan-2-boronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Benzofuran-2-boronic acid, 2-Benzofurylboronic acid

37873-0010 37873-0050 37873-0250
TS37873-0010EA 52 USD
TS37873-0010 TS37873-0050 TS37873-0250
Benzo[b]furan-2-boronic acid 97%
Benzo[b]furan-2-boronic acid
Formula: C₈H₇BO₃
Melting Pt: 112…114 °C
Storage Temperature: Ambient
MDL Number: MFCD00236019
CAS Number: 98437-24-2

Order Now

Specification Test Results

Appearance White to yellow Powder or crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR