Diethyl chlorophosphite 90% tech.

Supplier: Thermo Scientific Chemicals
Danger

11425-0250 11425-1000
TS11425-0250EA 122 USD
TS11425-0250 TS11425-1000
Diethyl chlorophosphite 90% tech.
Diethyl chlorophosphite
Formula: C₄H₁₀ClO₂P
Density: 1.09 g/cm³
Flash Pt: <1 °C
Storage Temperature: Ambient
MDL Number: MFCD00009074
CAS Number: 589-57-1
EINECS: 209-652-2
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4340 to 1.4410 (20°C, 589 nm)
Titration Argentometric ≥88 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR