4-Aminoantipyrine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Amino-2,3-dimethyl-1-phenyl-3-pyrazolin-5-one, Ampyrone, 4-Aminophenazone

10315-0025 10315-0250 10315-1000 10315-5000
TS10315-0025EA 1092 USD
TS10315-0025 TS10315-0250 TS10315-1000 TS10315-5000
4-Aminoantipyrine 98%
4-Aminoantipyrine
Formula: C₁₁H₁₃N₃O
MW: 203.24 g/mol
Melting Pt: 105.5…110 °C
Density: 0.8 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003145
CAS Number: 83-07-8
EINECS: 201-452-3
Merck Index: 13,00596

Order Now

Specification Test Results

Appearance Yellow to yellow-brown Powder or crystals
Infrared spectrum Conforms
Melting point 105 to 110 °C
Loss on drying ≤1.5 %
Sulfated ash ≤0.1 %
Solubility (5 % in water) Clear
Titration with HClO4 ≥97.5 % (On dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR