p-Toluenesulphonic acid monohydrate ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Methylbenzenesulphonic acid hydrate, Toluene-4-sulphonic acid monohydrate, 4-Methylbenzenesulfonic acid hydrate, Toluene-4-sulfonic acid monohydrate

42121-0250 42121-5000
TS42121-0250EA 29 USD
TS42121-0250 TS42121-1000 TS42121-5000
p-Toluenesulphonic acid monohydrate ACS
p-Toluenesulphonic acid monohydrate
Formula: C₇H₈O₃S·1H₂O
Melting Pt: 100…108 °C
Flash Pt: 180 °C
Storage Temperature: Ambient
MDL Number: MFCD00142137
CAS Number: 6192-52-5
UN: 2585
ADR: 8,III

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Water 9.5 to 11.5 %
Sulfated ash ≤0.1 %
Titration with NaOH ≥98.5 %
Clarity of solution (c=20, water) Passes test
Sulfate (SO4) ≤0.3 %
Iron (Fe) ≤0.01 %
Sodium (Na) ≤0.002 %
Heavy metals (as Pb) ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR