Picolinic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Pyridine-2-carboxylic acid, 2-Picolinic acid, 2-Pyridinecarboxylic acid
13117-0025
13117-0250
13117-1000
13117-5000
TS13117-0025EA
758
USD
TS13117-0025
TS13117-0250
TS13117-1000
TS13117-5000
Picolinic acid 99%
Picolinic acid
Formula:
C₆H₅NO₂ MW: 123.11 g/mol Melting Pt: 136…139 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006293 CAS Number: 98-98-6 EINECS: 202-719-7 Merck Index: 12,07557 |
Specification Test Results
Appearance | White to light yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 136 to 139 °C |
Titration with NaOH | 98.5 to 101.5 % |
Residue after evaporation | ≤0.001 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...