Methyl cinnamate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Methyl 3-phenylpropenoate, Methyl 3-phenyl-2-propenoate

27976-0050 27976-5000
TS27976-0050EA 31 USD
TS27976-0050 TS27976-5000
Methyl cinnamate 98%
Methyl cinnamate
Formula: C₁₀H₁₀O₂
MW: 162.19 g/mol
Melting Pt: 33.5…36 °C
Flash Pt: >110 °C
MDL Number: MFCD00008458
CAS Number: 103-26-4
EINECS: 203-093-8

Order Now

Specification Test Results

Appearance White to light yellow Fused crystalline mass
Infrared spectrum Conforms
Melting point 33.5 to 36 °C
GC ≥97.5 %
Acid value ≤1 mg KOH/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR