Formylhydrazine 90%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Formic acid hydrazide, Formohydrazide, Formic hydrazide

37606-0250 37606-1000
TS37606-0250EA 102 USD
TS37606-0250 TS37606-1000
Formylhydrazine 90%
Formylhydrazine
Formula: CH₄N₂O
MW: 60.06
Boiling Pt: 90
Melting Pt: 150
Flash Pt: 112 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007589
CAS Number: 624-84-0
EINECS: 210-867-9

Order Now

Specification Test Results

Appearance White to pink Crystalline powder or flakes
Infrared spectrum Conforms
Melting point 51 to 59 °C
Titration with KIO3 88 to 112 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR