1,5-Cyclooctadiene 99% stabilized
Supplier: Thermo Scientific Chemicals
|
Danger
|
29712-0010
29712-0025
29712-2500
TS29712-0010EA
119
USD
TS29712-0010
TS29712-0025
TS29712-2500
1,5-Cyclooctadiene 99% stabilized
1,5-Cyclooctadiene
Formula:
C₈H₁₂ Boiling Pt: 151 °C (101.3kPa) Melting Pt: –69.5 °C Density: 0.88 g/cm³ Flash Pt: 38 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001752 CAS Number: 111-78-4 EINECS: 203-907-1 UN: 2520 ADR: 3,III |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4920 to 1.4950 (20°C, 589 nm) |
GC | ≥98.5 % |
Stabilizer | -4'-hydroxyphenyl)propionate |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...