Ethyl oleate mixture of homologeous fatty acid esters 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Elaidic acid ethyl ester, (Z)-9-Octadecenoic acid ethyl ester, Oleic acid ethyl ester

41029-0010 41029-0025 41029-2500
TS41029-0010EA 114 USD
TS41029-0010 TS41029-0025 TS41029-2500
Ethyl oleate mixture of homologeous fatty acid esters 98%
Ethyl oleate
Formula: C₂₀H₃₈O₂
Boiling Pt: 190…210 °C (7 mmHg)
Density: 0.86 g/cm³
Flash Pt: 160 °C
Storage Temperature: Ambient
MDL Number: MFCD00009579
CAS Number: 111-62-6

Order Now

Specification Test Results

Appearance Clear viscous liquid
Infrared spectrum Conforms
Refractive index 1.4490 to 1.4530 (20°C, 589 nm)
Assay ≥97.5 % (ester)
Water ≤1 % (K.F.)
Density (20°C) 0.869 to 0.874
GC ≥60 % (oleic acid)
Color scale ≤70 APHA
Stabilizer 80 to 120 ppm BHA content
Acid value ≤0.5 mg KOH/g
Saponification value 177 to 188 mg KOH/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR