Methylphosphonic acid 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Methanephosphonic acid

35433-0050 35433-0250 35433-1000
TS35433-0050EA 77 USD
TS35433-0050 TS35433-0250 TS35433-1000
Methylphosphonic acid 98%
Methylphosphonic acid
Formula: CH₅O₃P
Melting Pt: 103…107 °C
Storage Temperature: Ambient
MDL Number: MFCD00002137
CAS Number: 993-13-5
EINECS: 213-607-2
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 103 to 107 °C
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR