Pyromellitic acid 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
10531-0050
10531-1000
10531-5000
TS10531-0050EA
24
USD
TS10531-0050
TS10531-1000
TS10531-5000
Pyromellitic acid 96%
Pyromellitic acid
Formula:
C₁₀H₆O₈ Boiling Pt: 516 °C (1013 hPa) Melting Pt: 281…284.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002471 CAS Number: 89-05-4 EINECS: 201-879-5 Merck Index: 12,08188 |
Specification Test Results
Appearance | White to light yellow Crystalline powder |
Infrared spectrum | Conforms |
Titration with NaOH | ≥95 % |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...