Teniposide
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(5R,5aR,8aR,9S)-9-(((2R,4aR,6R,7R,8R,8aS)-7,8-Dihydroxy-2-(thiophen-2-yl)hexahydropyrano[3,2-d][1,3]dioxin-6-yl)oxy)-5-(4-hydroxy-3,5-dimethoxyphenyl)-5,5a,8a,9-tetrahydrofuro[3',4':6,7]naphtho[2,3-d][1,3]dioxol-6(8H)-one
46685-2500
466850010
TS46685-2500EA
194
USD
TS46685-2500
TS466850010
Teniposide
Teniposide
Formula:
C₃₂H₃₂O₁₃S MW: 656.66 g/mol Melting Pt: 242°C…246°C Storage Temperature: Freezer |
MDL Number:
MFCD00866516 CAS Number: 29767-20-2 EINECS: 249-831-2 Merck Index: 13.9222 |
Specification Test Results
Appearance | White to almost white powder |
Infrared spectrum | Conforms |
HPLC | ≥ 96.0% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...