Adenosine-3',5'-cyclic monophosphate (cAMP) 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3',5'-cAMP, 3′,5′-Cyclic AMP, Adenosine 3′,5′-cyclophosphate, (4aR,6R,7R,7aS)-6-(6-Amino-9H-purin-9-yl)-2,7-dihydroxytetrahydro-4H-furo[3,2-d][1,3,2]dioxaphosphinine 2-oxide

22580-0010 22580-0050 22580-5000
TS22580-0010EA 199 USD
TS22580-0010 TS22580-0050 TS22580-5000
Adenosine-3',5'-cyclic monophosphate (cAMP) 99+%
Adenosine-3',5'-cyclic monophosphate (cAMP)
Formula: C₁₀H₁₂N₅O₆P
MW: 329.21 g/mol
Melting Pt: 260 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00005845
CAS Number: 60-92-4
UN: 3261
ADR: 8 III

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Water ≤7 % (K.F.)
HPLC ≥99.0 %
Ratio of Absorbances 0.13 to 0.19 (A280/A260)
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR