Ellagic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,3,7,8-Tetrahydroxychromeno[5,4,3-cde]chromene-5,10-dione, 4,4',5,5',6,6'-Hexahydroxydiphenic acid 2,6,2',6'-dilactone

11774-0100 11774-0025 11774-0500
TS11774-0100EA 217 USD
TS11774-0100 TS11774-0025 TS11774-0500
Ellagic acid 97%
Ellagic acid
Formula: C₁₄H₆O₈
MW: 302.194 g/mol
Melting Pt: >360 °C
Storage Temperature: Ambient
MDL Number: MFCD00006914
CAS Number: 476-66-4

Order Now

Specification Test Results

Appearance Beige to green Powder
Infrared spectrum Conforms
Titration with NaOH ≥96.0 %
Water ≤0.05 % (Coulometric)
GC ≥95 %
Color scale ≤10 APHA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR