Chloramphenicol sodium succinate 98-102%

Supplier: Thermo Scientific Chemicals
Warning

45953-0050 45953-0250
TS45953-0050EA 73 USD
TS45953-0050 TS45953-0250
Chloramphenicol sodium succinate 98-102%
Chloramphenicol sodium succinate
Formula: C₁₅H₁₅Cl₂N₂NaO₈
MW: 445.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00083598
CAS Number: 982-57-0

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Assay 98.0 to 102.0 % on dried substance
Water ≤2 %
Specific optical rotation +5.0° to +8.0° (20°C, 589 nm) (5 % in water) (on dried substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR