Vandetanib

Supplier: Thermo Scientific Chemicals

46433-2500 464330050 464330010
TS46433-2500EA 90 USD
TS46433-2500 TS464330050 TS464330010
Vandetanib
Vandetanib
Formula: C₂₂H₂₄BrFN₄O₂
Melting Pt: 240…243 °C
Storage Temperature: Freezer
MDL Number: MFCD07772346
CAS Number: 443913-73-3

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 %
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR