Glycitein
Supplier: Thermo Scientific Chemicals
Synonyms:
7-Hydroxy-3-(4-hydroxyphenyl)-6-methoxy-4H-chromen-4-one, 7,4'-Dihydroxy-6-methoxyisoflavone
46464-0500
464642500
TS46464-0500EA
211
USD
TS46464-0500
TS464642500
Glycitein
Glycitein
Formula:
C₁₆H₁₂O₅ MW: 284.27 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00016679 CAS Number: 40957-83-3 |
Specification Test Results
Appearance | White to light yellow Powder or crystals |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...