Ertapenem sodium salt 85%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ertapenem sodium, Sodium (4R,5S,6S)-3-(((3S,5S)-5-((3-carboxyphenyl)carbamoyl)pyrrolidin-3-yl)thio)-6-((R)-1-hydroxyethyl)-4-methyl-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylate

46064-0010 46064-2500
TS46064-0010EA 405 USD
TS46064-0010 TS46064-2500
Ertapenem sodium salt 85%
Ertapenem sodium salt
Formula: C₂₂H₂₄N₃NaO₇S
MW: 497.498 g/mol
Melting Pt: 230…234 °C
Storage Temperature: Freezer
MDL Number: MFCD09475566
CAS Number: 153773-82-1

Order Now

Specification Test Results

Appearance White to light yellow to light brown Powder
Infrared spectrum Conforms
Loss on drying 15 % to 19 %
HPLC ≥80 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR