2-Methyl-DL-serine hydrate 96%
Supplier: Thermo Scientific Chemicals
Synonyms:
2-Methylserine hydrate, (R,S)-2-Amino-3-hydroxy-2-methylpropionic acid hydrate
33975-0050
33975-0010
TS33975-0050EA
382
USD
TS33975-0050
TS33975-0010
2-Methyl-DL-serine hydrate 96%
2-Methyl-DL-serine hydrate
Formula:
C₄H₉NO₃.xH₂O MW: 119.12 g/mol (anhy) Melting Pt: 253…263 °C Storage Temperature: Ambient |
MDL Number:
MFCD03791288 CAS Number: 5424-29-3 EINECS: 226-565-5 |
Specification Test Results
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Water | ≤10 % (K.F.) |
Titration with HClO4 | ≥95 % (On dry substance) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...