Podophyllotoxin 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(10R,11R,15R,16R)-16-hydroxy-10-(3,4,5-trimethoxyphenyl)-4,6,13-trioxatetracyclo[7.7.0.03,7.011,15]hexadeca-1,3(7),8-trien-12-one, Condylox, PPT, (-)-Podophyllotoxin, Podofilox, Condyline, NSC 24818
22841-0010
22841-1000
TS22841-0010EA
443
USD
TS22841-0010
TS22841-1000
Podophyllotoxin 95%
Podophyllotoxin
Formula:
C₂₂H₂₂O₈ Melting Pt: 178…184 °C Storage Temperature: Freezer |
MDL Number:
MFCD00075290 CAS Number: 518-28-5 UN: 2811 ADR: 6.1,II |
Specification Test Results
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
HPLC | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...